Learn More
Selleck Chemical LLC LOX-IN-3 Dihydrochloride
SDP
Catalog No. 503137151
Encompass_Preferred
LOX-IN-3 Dihydrochloride is an orally active and selective lysyl oxidase (LOX) inhibitor
Catalog No. 50-313-7151 Supplier Selleck Chemical LLC Supplier No. E014525MG
LOX-IN-3 Dihydrochloride is an orally active and selective lysyl oxidase (LOX) inhibitor