Learn More
PHYTO AB INC LOX2 Antibody Immunogen: AT3G45140 Background: LOX2 (lipoxygenase 2) is a key enzyme in the JA biosynthetic pathway, and thereby regulates JA homeostasis in senescing leaves. 150ug/EA
Supplier:  PHYTO AB INC PHY2143S
Encompass_Preferred
PhytoAB Inc. is a contract research organization and plant antibody R&D based services and products supplier. PhytoAB is privately owned company headquatered in the San Francisco Bay Area of California, USA. PhytoAB specializes in antibody production for plant scientic usage and customized services, focus on the current need of plant antibodies to assist the further investigation of the current booming results from deep genome sequencing and increasing protomics studies. We have specialized R&D and customer services teams to fulfill the needs of botany & plant sciences research and industry world wide with high quality, professional and cutting edge services, and offering complete products usage guidance and suggestions for related problem solutions.
Catalog No. NC2877318