Learn More
Medchemexpress LLC Loxapine | 1977-10-2 | 327.81 | 100 MG
SDP
Catalog No. 5000385954
Encompass_Preferred
Loxapine (Standard) is the analytical standard of Loxapine, used for research and analytical applications. It functions as an orally active dopamine inhibitor, 5-HT receptor antagonist, and a dibenzoxazepine anti-psychotic agent. This analytical standard is commonly employed in qualitative, quantitative, and methodological research experiments involving HPLC, GC, and MS.
  • Used for research and analytical applications
  • Functions as a dopamine inhibitor
  • Acts as a 5-HT receptor antagonist
  • Used in HPLC, GC, and MS research experiments
Catalog No. 50-003-85954 Supplier Medchemexpress LLC Supplier No. HY17390100MG
Loxapine (Standard) is the analytical standard of Loxapine, used for research and analytical applications. It functions as an orally active dopamine inhibitor, 5-HT receptor antagonist, and a dibenzoxazepine anti-psychotic agent. This analytical standard is commonly employed in qualitative, quantitative, and methodological research experiments involving HPLC, GC, and MS.
  • Used for research and analytical applications
  • Functions as a dopamine inhibitor
  • Acts as a 5-HT receptor antagonist
  • Used in HPLC, GC, and MS research experiments