Learn More
Cayman Chemical Loxapine (succinate), Purity: ≥98%, CAS: 27833-64-3, 50 mg
SDP
Catalog No. 502983354
Encompass_Preferred
An atypical antipsychotic drug that displays high affinity for histamine (H), serotonin (5-HT), dopamine (D), and α1-adrenergic receptors (Ki values are 7, 7.7, 9.5, 12, and 31 nM for H1, 5-HT2A, 5-HT2C, D2, and α1A, respectively)
Catalog No. 50-298-3354 Supplier Cayman Chemical Supplier No. 2076050 MG
An atypical antipsychotic drug that displays high affinity for histamine (H), serotonin (5-HT), dopamine (D), and α1-adrenergic receptors (Ki values are 7, 7.7, 9.5, 12, and 31 nM for H1, 5-HT2A, 5-HT2C, D2, and α1A, respectively)