Learn More
Medchemexpress LLC HY-B2154 10mg Medchemexpress, Loxiglumide CAS:107097-80-3 Purity:>98%
SDP
Supplier:  Medchemexpress LLC HYB215410MG
Encompass_Preferred
Medchemexpress, HY-B2154 10mg Loxiglumide CAS:107097-80-3 Loxiglumide is a cholecystokinin ( CCK-1 ) receptor antagonist. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-193-3086