Learn More
Selleck Chemical LLC Loxistatin Acid
SDP
Catalog No. 5000004233
Encompass_Preferred
Loxistatin Acid (E-64C NSC 694279 EP 475) a derivative of E-64 is an irreversible and membrane-permeant cysteine protease inhibitor The cysteine protease cathepsin L is required for SARS-CoV-2 viral entry
Catalog No. 50-000-04233 Supplier Selleck Chemical LLC Supplier No. S739225MG
Loxistatin Acid (E-64C NSC 694279 EP 475) a derivative of E-64 is an irreversible and membrane-permeant cysteine protease inhibitor The cysteine protease cathepsin L is required for SARS-CoV-2 viral entry