Learn More
Medchemexpress LLC Loxistatin acid | 76684-89-4 | 99.3% | 314.38 g/mol | C15H26N2O5 | 5 MG
SDP
Catalog No. 5000215754
Encompass_Preferred
E 64c (loxistatin acid) is a cysteine protease inhibitor used in research to inhibit calpain and related cysteine proteases in biochemical and cell-based assays. Supplied as a high-purity small-molecule solid, it is intended for enzymology, protease inhibition studies, and cellular investigations.
  • High purity (as supplied) suitable for analytical and biological assays.
  • Irreversible inhibitor of cysteine proteases including calpain.
  • Solid form with good solubility in DMSO for stock solution preparation.
  • Applicable to biochemical, enzymatic, and cell-based studies of protease function.
  • Documentation available including data sheet and certificate of analysis.
Catalog No. 50-002-15754 Supplier Medchemexpress LLC Supplier No. HY1002275MG
E 64c (loxistatin acid) is a cysteine protease inhibitor used in research to inhibit calpain and related cysteine proteases in biochemical and cell-based assays. Supplied as a high-purity small-molecule solid, it is intended for enzymology, protease inhibition studies, and cellular investigations.
  • High purity (as supplied) suitable for analytical and biological assays.
  • Irreversible inhibitor of cysteine proteases including calpain.
  • Solid form with good solubility in DMSO for stock solution preparation.
  • Applicable to biochemical, enzymatic, and cell-based studies of protease function.
  • Documentation available including data sheet and certificate of analysis.