Learn More
Selleck Chemical LLC Loxistatin Acid (E-64C) 5mg 76684-89-4 NSC 694279, EP 475
SDP
Catalog No. NC1851128
Encompass_Preferred
Loxistatin Acid (E-64C, NSC 694279, EP 475), a derivative of E-64, is an irreversible and membrane-permeant cysteine protease inhibitor. The cysteine protease cathepsin L is required for SARS-CoV-2 viral entry. *For Research & Development use only.  Product is not intended for drug, household, or human consumption.
Catalog No. NC1851128 Supplier Selleck Chemical LLC Supplier No. S73925MG
Loxistatin Acid (E-64C, NSC 694279, EP 475), a derivative of E-64, is an irreversible and membrane-permeant cysteine protease inhibitor. The cysteine protease cathepsin L is required for SARS-CoV-2 viral entry. *For Research & Development use only.  Product is not intended for drug, household, or human consumption.