Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Loxistatin acid (E-64c) | 76684-89-4 | 99.3% | 314.38 | C15H26N2O5 | 25 MG
SDP

Catalog No. 5000215589
Encompass_Preferred

E 64c (Loxistatin acid) is an irreversible inhibitor of cysteine proteases derived from the natural epoxide E-64. It is intended for research use in biochemical and enzymology assays to study calpain and other cysteine proteases. The material is supplied as a high-purity white to off-white solid and is available in multiple package sizes and preprepared DMSO solution formats for immediate use in assays.

  • Irreversible cysteine protease inhibitor
  • High purity (99.32%)
  • Molecular weight 314.38 g·mol⁻1
  • Available as solid and as 10 mM solution in DMSO
  • Suitable for calpain and protease inhibition assays
  • Storage conditions provided for powder and solvent formats

Catalog No. 50-002-15589 Supplier Medchemexpress LLC Supplier No. HY10022725MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

E 64c (Loxistatin acid) is an irreversible inhibitor of cysteine proteases derived from the natural epoxide E-64. It is intended for research use in biochemical and enzymology assays to study calpain and other cysteine proteases. The material is supplied as a high-purity white to off-white solid and is available in multiple package sizes and preprepared DMSO solution formats for immediate use in assays.

  • Irreversible cysteine protease inhibitor
  • High purity (99.32%)
  • Molecular weight 314.38 g·mol⁻1
  • Available as solid and as 10 mM solution in DMSO
  • Suitable for calpain and protease inhibition assays
  • Storage conditions provided for powder and solvent formats

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.