Learn More
Medchemexpress LLC Loxistatin acid (E-64c) | 76684-89-4 | MFCD00132882 | 99.3% | 314.38 g/mol | C15H26N2O5 | 10 MG
SDP
Catalog No. 5000215601
Encompass_Preferred
Loxistatin acid (E-64c) is an irreversible epoxide-based inhibitor of cysteine proteases used as a research reagent to probe protease-dependent biological processes. It is a membrane-permeant derivative of the natural epoxide inhibitor E-64, with reported activity against papain-like enzymes, cathepsins, and calcium-activated neutral protease; it has also shown entry-blocking effects in MERS-CoV studies.
  • Irreversible epoxide inhibitor of cysteine proteases.
  • Membrane-permeant chemistry suitable for cell-based assays.
  • Reported activity against papain, cathepsins, and CANP.
  • High purity for reproducible experimental results.
  • Recommended storage preserves stability for long-term use.
Catalog No. 50-002-15601 Supplier Medchemexpress LLC Supplier No. HY10022710MG
Loxistatin acid (E-64c) is an irreversible epoxide-based inhibitor of cysteine proteases used as a research reagent to probe protease-dependent biological processes. It is a membrane-permeant derivative of the natural epoxide inhibitor E-64, with reported activity against papain-like enzymes, cathepsins, and calcium-activated neutral protease; it has also shown entry-blocking effects in MERS-CoV studies.
  • Irreversible epoxide inhibitor of cysteine proteases.
  • Membrane-permeant chemistry suitable for cell-based assays.
  • Reported activity against papain, cathepsins, and CANP.
  • High purity for reproducible experimental results.
  • Recommended storage preserves stability for long-term use.