Learn More
Aviva Systems Biology Corp LOXL1 Antibody - middle region (ARP54781_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP54781_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against LOXL1. It was validated on Western Blot using a cell lysate as a positive control.The immunogen is a synthetic peptide directed towards the middle region of human LOXL1. Alternate names: LOL, LOXLApplicationWB. Species reactivity: Cow, Guinea Pig, Horse, Human, Mouse, Pig, Rabbit, Rat, Zebrafish
Catalog No. 50-014-61312