Learn More
ACROBiosystems ACROBiosystems Biotinylated Human LOXL2 Protein, His,Avitag™
Catalog No. 501956978
Encompass_Preferred
Recombinant Protein;25UG;Biotinylated Human LOXL2, His,Avitag (LO2-H82E3) is expressed from human 293 cells (HEK293). It contains AA Gln 26 - Gln 774 (Accession # Q9Y4K0-1).;This protein carries a polyhistidine tag at the C-terminus, followed by an Avi tag. The protein has a calculated MW of 87.8 kDa.
Catalog No. 50-195-6978 Supplier ACROBiosystems Supplier No. LO2H82E325UG
Recombinant Protein;25UG;Biotinylated Human LOXL2, His,Avitag (LO2-H82E3) is expressed from human 293 cells (HEK293). It contains AA Gln 26 - Gln 774 (Accession # Q9Y4K0-1).;This protein carries a polyhistidine tag at the C-terminus, followed by an Avi tag. The protein has a calculated MW of 87.8 kDa.