Learn More
Apexbio Technology LLC LOXO-101 1223405-08-0 5mg
SDP
Catalog No. 5000092133
Encompass_Preferred
LOXO-101 (CAS 1223405-08-0) is a potent and selective inhibitor of the tropomyosin receptor kinase (TRK) family demonstrating low nanomolar inhibitory activity across all TRK isoforms LOXO-101 disrupts signaling mediated by neurotrophin receptors which are critical for neuronal development and survival In kinase profiling LOXO-101 exhibited minimal off-target activity with significant inhibition observed only for TNK2 (IC50 576 nM) and did not display hERG inhibition or QT prolongation effects In vivo studies revealed dose-dependent tumor growth suppression in xenograft models Clinically LOXO-101 has been evaluated for safety and pharmacokinetics and induced rapid tumor regression in a TRK fusion-positive patient highlighting its utility in studying TRK-driven malignancies
Catalog No. 50-000-92133 Supplier Apexbio Technology LLC Supplier No. B61765
LOXO-101 (CAS 1223405-08-0) is a potent and selective inhibitor of the tropomyosin receptor kinase (TRK) family demonstrating low nanomolar inhibitory activity across all TRK isoforms LOXO-101 disrupts signaling mediated by neurotrophin receptors which are critical for neuronal development and survival In kinase profiling LOXO-101 exhibited minimal off-target activity with significant inhibition observed only for TNK2 (IC50 576 nM) and did not display hERG inhibition or QT prolongation effects In vivo studies revealed dose-dependent tumor growth suppression in xenograft models Clinically LOXO-101 has been evaluated for safety and pharmacokinetics and induced rapid tumor regression in a TRK fusion-positive patient highlighting its utility in studying TRK-driven malignancies