Learn More
Abcam LO x O-101, 5MG
Supplier:  Abcam AB2869905MG
Encompass_Preferred
MW 428.4 Da, Purity >=98%. Potent, ATP-competitive TRK inhibitor with IC₅₀s in low nanomolar range for inhibition of all TRK family members in binding and cellular assays, with 100-fold selectivity over other kinases, with 2 to 20 nM cellular potency against the TRKA, TRKB, and TRKC kinases.
The product is subject to the following: Abcam Restricted Use Statement
Catalog No. 50-220-5859