Learn More
Medchemexpress LLC HY-B0578 10mg Medchemexpress, Loxoprofen CAS:68767-14-6 Purity:>98%
SDP
Catalog No. 502082608
Encompass_Preferred
Medchemexpress, HY-B0578 10mg Loxoprofen CAS:68767-14-6 Loxoprofen is a non-steroidal anti-inflammatory agent with analgesic and anti-pyretic properties. Loxoprofen sodium is a nonselective COX inhibitor with IC50s of 6.5 and 13.5 μM for COX-1 and COX-2, respectively. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-208-2608 Supplier Medchemexpress LLC Supplier No. HYB057810MG
Medchemexpress, HY-B0578 10mg Loxoprofen CAS:68767-14-6 Loxoprofen is a non-steroidal anti-inflammatory agent with analgesic and anti-pyretic properties. Loxoprofen sodium is a nonselective COX inhibitor with IC50s of 6.5 and 13.5 μM for COX-1 and COX-2, respectively. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.