Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Loxoprofen L-menthol ester | 1091621-63-4 | 99.7% | 384.55 | C25H36O3 | 10 MG
SDP

Catalog No. 5000208400
Encompass_Preferred

Loxoprofen L-menthol ester is a degradation product of loxoprofen sodium intended for research and analytical studies. The compound has a molecular formula of C25H36O3, a molecular weight of 384.55, and is supplied as a small research sample for degradation or analytical work.

  • High purity: 99.73% (as reported)
  • Appearance: oil; white to off-white color
  • Storage: 4°C, protect from light
  • In-solvent stability: -80°C for 6 months; -20°C for 1 month (protect from light)
  • Supplied as a 10 mg research quantity

Catalog No. 50-002-08400 Supplier Medchemexpress LLC Supplier No. HYW08720710MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Loxoprofen L-menthol ester is a degradation product of loxoprofen sodium intended for research and analytical studies. The compound has a molecular formula of C25H36O3, a molecular weight of 384.55, and is supplied as a small research sample for degradation or analytical work.

  • High purity: 99.73% (as reported)
  • Appearance: oil; white to off-white color
  • Storage: 4°C, protect from light
  • In-solvent stability: -80°C for 6 months; -20°C for 1 month (protect from light)
  • Supplied as a 10 mg research quantity

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.