Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Loxoprofen L-menthol ester | 1091621-63-4 | 99.7% | 384.55 | C25H36O3 | 100 MG
SDP

Catalog No. 5000208540
Encompass_Preferred

Loxoprofen L-menthol ester is the menthol ester degradation product of loxoprofen supplied for research use. It is a white to off-white oil with defined molecular properties and high analytical purity.

  • Degradation product of loxoprofen.
  • CAS number 1091621-63-4.
  • Molecular formula C25H36O3.
  • Molecular weight 384.55 g/mol.
  • High purity (99.7%).
  • Available in 100 MG pack size.

Catalog No. 50-002-08540 Supplier Medchemexpress LLC Supplier No. HYW087207100MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Loxoprofen L-menthol ester is the menthol ester degradation product of loxoprofen supplied for research use. It is a white to off-white oil with defined molecular properties and high analytical purity.

  • Degradation product of loxoprofen.
  • CAS number 1091621-63-4.
  • Molecular formula C25H36O3.
  • Molecular weight 384.55 g/mol.
  • High purity (99.7%).
  • Available in 100 MG pack size.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.