Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Loxoprofen L-menthol ester | 1091621-63-4 | 99.7% | 384.55 | C25H36O3 | 5 MG
SDP

Catalog No. 5000208539
Encompass_Preferred

Loxoprofen L-menthol ester is a chemical degradation product of loxoprofen sodium, supplied as a research reagent for analytical and stability studies. It is used as a reference standard or analytical target in pharmaceutical research.

  • Chemical degradation product of loxoprofen sodium.
  • High purity suitable for analytical use (purity 99.7%).
  • CAS number 1091621-63-4.
  • Molecular formula C25H36O3; molecular weight 384.55 g/mol.
  • Available in small research sizes, such as 5 mg.

Catalog No. 50-002-08539 Supplier Medchemexpress LLC Supplier No. HYW0872075MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Loxoprofen L-menthol ester is a chemical degradation product of loxoprofen sodium, supplied as a research reagent for analytical and stability studies. It is used as a reference standard or analytical target in pharmaceutical research.

  • Chemical degradation product of loxoprofen sodium.
  • High purity suitable for analytical use (purity 99.7%).
  • CAS number 1091621-63-4.
  • Molecular formula C25H36O3; molecular weight 384.55 g/mol.
  • Available in small research sizes, such as 5 mg.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.