Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Loxoprofen L-menthol ester | 1091621-63-4 | 99.7% | 384.55 g/mol | C25H36O3 | 25 MG
SDP

Catalog No. 5000208644
Encompass_Preferred

Loxoprofen L-menthol ester is a degradation product of loxoprofen sodium supplied for research and analytical use. It is intended for use in stability studies, method development, and as an analytical reference standard.

  • High purity suitable for analytical use.
  • Provided in milligram-scale quantities for laboratory work.
  • Well characterized with CAS 1091621-63-4 and formula C25H36O3.
  • Useful as a degradation-product standard in stability testing.

Catalog No. 50-002-08644 Supplier Medchemexpress LLC Supplier No. 25MGHYW087207
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Loxoprofen L-menthol ester is a degradation product of loxoprofen sodium supplied for research and analytical use. It is intended for use in stability studies, method development, and as an analytical reference standard.

  • High purity suitable for analytical use.
  • Provided in milligram-scale quantities for laboratory work.
  • Well characterized with CAS 1091621-63-4 and formula C25H36O3.
  • Useful as a degradation-product standard in stability testing.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.