Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Loxoprofen L-menthol ester | 1091621-63-4 | 99.7% | 384.55 g/mol | C25H36O3 | 50 MG
SDP

Catalog No. 5000208359
Encompass_Preferred

Loxoprofen L-menthol ester is a chemical degradation product of loxoprofen sodium provided for laboratory research and analytical applications; it is related to non-steroidal anti-inflammatory agents and supplied at high purity for experimental use.

  • Chemical degradation product of loxoprofen sodium.
  • Intended for laboratory research and analytical applications.
  • High purity (≈99.7%).
  • Appearance reported as oil, white to off-white.
  • Storage: 4°C, protect from light; in solvent: -80°C (up to 6 months) or -20°C (up to 1 month).

Catalog No. 50-002-08359 Supplier Medchemexpress LLC Supplier No. HYW08720750MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Loxoprofen L-menthol ester is a chemical degradation product of loxoprofen sodium provided for laboratory research and analytical applications; it is related to non-steroidal anti-inflammatory agents and supplied at high purity for experimental use.

  • Chemical degradation product of loxoprofen sodium.
  • Intended for laboratory research and analytical applications.
  • High purity (≈99.7%).
  • Appearance reported as oil, white to off-white.
  • Storage: 4°C, protect from light; in solvent: -80°C (up to 6 months) or -20°C (up to 1 month).

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.