Learn More
Medchemexpress LLC Loxoprofen sodium | 80382-23-6 | 99.6% | 1 ML
SDP
Catalog No. 5000409034
Encompass_Preferred
Loxoprofen sodium is a non-steroidal, orally active anti-inflammatory agent with analgesic and anti-pyretic properties. It is a nonselective COX inhibitor with IC50s of 6.5 and 13.5 μM for COX-1 and COX-2, respectively. It can also reduce atherosclerosis and shows antitumor activity.
  • Non-steroidal, orally active anti-inflammatory agent
  • Analgesic and anti-pyretic properties
  • Nonselective COX inhibitor
  • Reduces atherosclerosis
  • Shows antitumor activity
Catalog No. 50-004-09034 Supplier Medchemexpress LLC Supplier No. HYB0578A10MM/1ML
Loxoprofen sodium is a non-steroidal, orally active anti-inflammatory agent with analgesic and anti-pyretic properties. It is a nonselective COX inhibitor with IC50s of 6.5 and 13.5 μM for COX-1 and COX-2, respectively. It can also reduce atherosclerosis and shows antitumor activity.
  • Non-steroidal, orally active anti-inflammatory agent
  • Analgesic and anti-pyretic properties
  • Nonselective COX inhibitor
  • Reduces atherosclerosis
  • Shows antitumor activity