Learn More
Cayman Chemical Loxoprofen-SRS, Purity: ≥98%, CAS: 83648-76-4, 10 mg
SDP
Catalog No. 502916570
Encompass_Preferred
An NSAID, a COX inhibitor (IC50s = 0.5 and 0.39 for recombinant human COX-1 and COX-2, respectively), and an active metabolite of loxoprofen; intravenous administration inhibits acetic acid-induced writhing, reduces carrageenan-induced mechanical hyperalgesia, and decreases carrageenan-induced paw edema in rats (ED50s = 0.43, 1.28, and 8.01 mg/kg, respectively)
Catalog No. 50-291-6570 Supplier Cayman Chemical Supplier No. 3812210 MG
An NSAID, a COX inhibitor (IC50s = 0.5 and 0.39 for recombinant human COX-1 and COX-2, respectively), and an active metabolite of loxoprofen; intravenous administration inhibits acetic acid-induced writhing, reduces carrageenan-induced mechanical hyperalgesia, and decreases carrageenan-induced paw edema in rats (ED50s = 0.43, 1.28, and 8.01 mg/kg, respectively)