Learn More
Cayman Chemical Loxoprofen (sodium salt), Purity: ≥98%, CAS: 80382-23-6, 5 g
SDP
Catalog No. 502911530
Encompass_Preferred
An NSAID and a prodrug form of loxoprofen-SRS; inhibits COX-1 and COX-2 in isolated human whole blood (IC50s = 6.5 and 13.5 µM, respectively); reduces acetic acid-induced writhing in mice and carrageenan-induced paw edema and LPS-induced fever in rats (ED30s = 20.1, 0.7, and 2.79 mg/kg, respectively)
Catalog No. 50-291-1530 Supplier Cayman Chemical Supplier No. 350885 G
An NSAID and a prodrug form of loxoprofen-SRS; inhibits COX-1 and COX-2 in isolated human whole blood (IC50s = 6.5 and 13.5 µM, respectively); reduces acetic acid-induced writhing in mice and carrageenan-induced paw edema and LPS-induced fever in rats (ED30s = 20.1, 0.7, and 2.79 mg/kg, respectively)