Learn More
Medchemexpress LLC HY-108472 10mg Medchemexpress, Loxoribine CAS:121288-39-9 Purity:>98%
SDP
Catalog No. 501968555
Encompass_Preferred
Medchemexpress, HY-108472 10mg Loxoribine CAS:121288-39-9 Loxoribine (7-Allyl-8-oxoguanosine) is a guanosine analog with anti-viral and anti-tumor activities. Loxoribine is an orally bioavailable and selective Toll-like receptor (TLR) 7 agonist[3]. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-196-8555 Supplier Medchemexpress LLC Supplier No. HY10847210MG
Medchemexpress, HY-108472 10mg Loxoribine CAS:121288-39-9 Loxoribine (7-Allyl-8-oxoguanosine) is a guanosine analog with anti-viral and anti-tumor activities. Loxoribine is an orally bioavailable and selective Toll-like receptor (TLR) 7 agonist[3]. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.