Learn More
Enzo Life Sciences Loxoribine (25mg). CAS: 121288-39-9
SDP
Supplier:  Enzo Life Sciences ALX480097M025
Encompass_Preferred
Selective ligand for Toll-like receptor 7 (TLR7) via IFN. Activates natural killer cells and primes cytolytic precursor cells for activation by IL-2. Alternative name: 7-Allyl-8-oxoguanosine, RWJ 21757. Purity: ≥97% (HPLC (UV)). Appearance: White to off-white solid. Solubility: Soluble in DMSO (30mg/ml), water (10mg/ml), water:acetonitrile (1:1) (20mg/ml) or water:methanol (1:1) (10mg/ml). Long Term Storage: -20°C.
Catalog No. 50-200-9327