Learn More
AdipoGen Loxoribine (25 mg)
SDP
Supplier:  AdipoGen AGCR13584M025
Encompass_Preferred
Chemical. CAS: 121288-39-9. Formula: C13H17N5O6. MW: 339.3. Guanosine analog derivatized at position N7 and C8. Very potent immunostimulatory compound that increases cytokine production. Antitumor and antiviral activity compound. Potent and selective synthetic ligand for Toll-like receptor 7 (TLR7). Co-incubation with thymidine homopolymer oligodeoxynucleotides (ODN) redirected the stimulatory effect of loxoribine away from TLR7 toward TLR8. Induces autophagy and autophagy-induced cell death in Caco-2 cells. Shows adjuvant activity to increase the effectiveness of vaccines.
Catalog No. 50-205-3951