Learn More
Medchemexpress LLC HY-111455 5mg Medchemexpress, LP-211 CAS:1052147-86-0 Purity:>98%
SDP
Catalog No. 501968874
Encompass_Preferred
Medchemexpress, HY-111455 5mg LP-211 CAS:1052147-86-0 LP-211 is a selective and blood−brain barrier penetrant 5-HT7 receptor agonist, with a Ki of 0.58 nM, with high selectivity over 5-HT1A receptor (Ki, 188 nM) and D2 receptor (Ki, 142 nM). Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-196-8874 Supplier Medchemexpress LLC Supplier No. HY1114555MG
Medchemexpress, HY-111455 5mg LP-211 CAS:1052147-86-0 LP-211 is a selective and blood−brain barrier penetrant 5-HT7 receptor agonist, with a Ki of 0.58 nM, with high selectivity over 5-HT1A receptor (Ki, 188 nM) and D2 receptor (Ki, 142 nM). Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.