Learn More
Miele, Inc. LP 2800 Cartridge Empty 19L. R
Catalog No. 502870711
Encompass_Preferred
LP 2800 Cartridge Empty 19L. Resin capacity: 5,600L at 150µs/cm H570 Ø230mm
Catalog No. 50-287-0711 Supplier Miele, Inc. Supplier No. 9782590
LP 2800 Cartridge Empty 19L. Resin capacity: 5,600L at 150µs/cm H570 Ø230mm