Learn More
TARGETMOL CHEMICALS INC LP-471756 5MG
SDP
Catalog No. 502580563
Encompass_Preferred
Also available in 10 mg 25 mg 50 mg 100 mg 200 mg 500 mg 1 mL 10 mM (in DMSO) and bulk. Please contact Fisher for quotes. LP-471756 is a specific antagonist of GPR139 and inhibits LP-360924-stimulated cAMP production (IC50 = 640 nM). purity: 99%
Catalog No. 50-258-0563 Supplier TARGETMOL CHEMICALS INC Supplier No. T278505MG
Also available in 10 mg 25 mg 50 mg 100 mg 200 mg 500 mg 1 mL 10 mM (in DMSO) and bulk. Please contact Fisher for quotes. LP-471756 is a specific antagonist of GPR139 and inhibits LP-360924-stimulated cAMP production (IC50 = 640 nM). purity: 99%