Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC LP-922761 hydrate | 00-00-0 | 98.0% | 414.98 | C21H26N6O3.1/4H2O | 5 MG
SDP

Catalog No. 5000262060
Encompass_Preferred

LP-922761 hydrate is a potent, selective, orally active inhibitor of adapter protein-2 associated kinase 1 (AAK1) intended for research use. It shows low-nanomolar potency in biochemical and cellular assays and also inhibits BMP-2-inducible kinase (BIKE). The compound is supplied as a solid hydrate with high purity and requires cold storage for long-term stability.

  • Potent AAK1 inhibition with IC50 4.8 nM (enzyme) and 7.6 nM (cell).
  • Secondary activity against BIKE (IC50 24 nM).
  • High purity (98.0%) suitable for research applications.
  • Appearance: white to off-white solid.
  • Storage: powder -20°C for up to 3 years; in solvent keep frozen for shorter terms.
  • For research use only; not for human or therapeutic use.

Catalog No. 50-002-62060 Supplier Medchemexpress LLC Supplier No. HY120179A5MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

LP-922761 hydrate is a potent, selective, orally active inhibitor of adapter protein-2 associated kinase 1 (AAK1) intended for research use. It shows low-nanomolar potency in biochemical and cellular assays and also inhibits BMP-2-inducible kinase (BIKE). The compound is supplied as a solid hydrate with high purity and requires cold storage for long-term stability.

  • Potent AAK1 inhibition with IC50 4.8 nM (enzyme) and 7.6 nM (cell).
  • Secondary activity against BIKE (IC50 24 nM).
  • High purity (98.0%) suitable for research applications.
  • Appearance: white to off-white solid.
  • Storage: powder -20°C for up to 3 years; in solvent keep frozen for shorter terms.
  • For research use only; not for human or therapeutic use.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.