Learn More
Medchemexpress LLC HY-117626 5mg Medchemexpress, LP-935509 CAS:1454555-29-3 Purity:>98%
SDP
Catalog No. 502029831
Encompass_Preferred
Medchemexpress, HY-117626 5mg LP-935509 CAS:1454555-29-3 LP-935509 is a selective, ATP-competitive and brain-penetrant inhibitor of adapter protein-2 associated kinase 1 (AAK1) with an IC50 and a Ki of 3.3 nM and 0.9 nM, respectively. LP-935509 is also a potent inhibitor of BIKE (IC50=14 nM) and a modest inhibitor of GAK (IC50=320 nM). LP-935509 is highly brain-penetrant and reverses fully established pain behavior following the SNL procedure. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-202-9831 Supplier Medchemexpress LLC Supplier No. HY1176265MG
Medchemexpress, HY-117626 5mg LP-935509 CAS:1454555-29-3 LP-935509 is a selective, ATP-competitive and brain-penetrant inhibitor of adapter protein-2 associated kinase 1 (AAK1) with an IC50 and a Ki of 3.3 nM and 0.9 nM, respectively. LP-935509 is also a potent inhibitor of BIKE (IC50=14 nM) and a modest inhibitor of GAK (IC50=320 nM). LP-935509 is highly brain-penetrant and reverses fully established pain behavior following the SNL procedure. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.