Learn More
Randox LP(A) CALIBRATOR SERIES
SDP
Catalog No. NC0010821
Encompass_Preferred
Lipoprotein (a) Calibrator; 5-point calibrator which takes into account the heterogeneity of the Lp(a) molecule for each of the levels, resulting in excellent commutability of the calibrator with patient samples
Catalog No. NC0010821 Supplier Randox Supplier No. LP3404
Lipoprotein (a) Calibrator; 5-point calibrator which takes into account the heterogeneity of the Lp(a) molecule for each of the levels, resulting in excellent commutability of the calibrator with patient samples