Learn More
WELCH MATERIALS INC Ultisil LP-AQ, 5µm, 120Å, 4.6×50mm
Catalog No. NC2052215
Encompass_Preferred
Analytical Column, Ultisil LP-AQ, 5μm, 120Å, 4.6×50mm, pH range: 1.0-8.0, Pore Size: 120Å, USP listing: L1, Carbon Loading: 5%, Surface Area: 320m2/g, not-endcapped.
Catalog No. NC2052215 Supplier WELCH MATERIALS INC Supplier No. H0025931037
Analytical Column, Ultisil LP-AQ, 5μm, 120Å, 4.6×50mm, pH range: 1.0-8.0, Pore Size: 120Å, USP listing: L1, Carbon Loading: 5%, Surface Area: 320m2/g, not-endcapped.