Learn More
WELCH MATERIALS INC Ultisil LP-C18, 10µm, 50×250mm
Catalog No. NC2052609
Encompass_Preferred
Prep Column, Ultisil LP-C18, 10µm, 50×250mm, pH range: 0.5-8.0, Pore Size: 120Å, USP listing: L1, Carbon Loading: 10%, Surface Area: 320m2/g, not-endcapped.
Catalog No. NC2052609 Supplier WELCH MATERIALS INC Supplier No. H0261331210
Add to Cart
Add to Cart
Request a Quote
Prep Column, Ultisil LP-C18, 10µm, 50×250mm, pH range: 0.5-8.0, Pore Size: 120Å, USP listing: L1, Carbon Loading: 10%, Surface Area: 320m2/g, not-endcapped.