Learn More
WELCH MATERIALS INC Ultisil LP-C18, 5µm, 21.2×250mm
Supplier:  WELCH MATERIALS INC H0261321106
Encompass_Preferred
Prep Column, Ultisil LP-C18, 5µm, 21.2×250mm, pH range: 0.5-8.0, Pore Size: 120Å, USP listing: L1, Carbon Loading: 10%, Surface Area: 320m2/g, not-endcapped.
Catalog No. NC2052606