Learn More
WELCH MATERIALS INC Ultisil LP-C18, 5µm,300Å, 4.6×250mm
Catalog No. NC2052218
Encompass_Preferred
Analytical Column, Ultisil LP-C18, 5μm, 300Å, 4.6×250mm, pH range: 0.5-8.0, Pore Size: 300Å, USP listing: L1, Carbon Loading: 5%, Surface Area: 90m2/g, not-endcapped.
Catalog No. NC2052218 Supplier WELCH MATERIALS INC Supplier No. H0020833043
Analytical Column, Ultisil LP-C18, 5μm, 300Å, 4.6×250mm, pH range: 0.5-8.0, Pore Size: 300Å, USP listing: L1, Carbon Loading: 5%, Surface Area: 90m2/g, not-endcapped.