Learn More
Medchemexpress LLC HY-19757 5mg Medchemexpress, Lp-PLA2-IN-1 CAS:1420367-28-7 Purity:>98%
SDP
Catalog No. 502082177
Encompass_Preferred
Medchemexpress, HY-19757 5mg Lp-PLA2-IN-1 CAS:1420367-28-7 Lp-PLA2-IN-1 is a potent Lipoprotein-associated phospholipase A2 (Lp-PLA2) inhibitor. Lp-PLA2-IN-1 has the potential for atherosclerosis, Alzheimer's disease research. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-208-2177 Supplier Medchemexpress LLC Supplier No. HY197575MG
Medchemexpress, HY-19757 5mg Lp-PLA2-IN-1 CAS:1420367-28-7 Lp-PLA2-IN-1 is a potent Lipoprotein-associated phospholipase A2 (Lp-PLA2) inhibitor. Lp-PLA2-IN-1 has the potential for atherosclerosis, Alzheimer's disease research. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.