Learn More
Medchemexpress LLC Lp-PLA2-IN-3 | 2196245-16-4 | 99.9% | 467.85 g/mol | C20H13ClF3N3O3S | 5 MG
SDP
Catalog No. 5000262227
Encompass_Preferred
Lp-PLA2-IN-3 is a potent, orally bioavailable small-molecule inhibitor of lipoprotein-associated phospholipase A2 supplied for research use. It inhibits recombinant human Lp-PLA2 with an IC50 of 14 nM and is provided as a white to off-white solid suitable for in vitro and in vivo studies.
  • Potent inhibition of Lp-PLA2 with an IC50 of 14 nM.
  • High purity (99.9%) for reliable experimental results.
  • Orally bioavailable profile suitable for pharmacology studies.
  • Soluble at ≥ 250 mg/mL in DMSO and formulatable for in vivo dosing.
  • Stable as a powder when stored at -20°C for long-term storage.
Catalog No. 50-002-62227 Supplier Medchemexpress LLC Supplier No. HY1331495MG
Lp-PLA2-IN-3 is a potent, orally bioavailable small-molecule inhibitor of lipoprotein-associated phospholipase A2 supplied for research use. It inhibits recombinant human Lp-PLA2 with an IC50 of 14 nM and is provided as a white to off-white solid suitable for in vitro and in vivo studies.
  • Potent inhibition of Lp-PLA2 with an IC50 of 14 nM.
  • High purity (99.9%) for reliable experimental results.
  • Orally bioavailable profile suitable for pharmacology studies.
  • Soluble at ≥ 250 mg/mL in DMSO and formulatable for in vivo dosing.
  • Stable as a powder when stored at -20°C for long-term storage.