Learn More
Medchemexpress LLC Lp-PLA2-IN-3 | 2196245-16-4 | 99.9% | C20H13ClF3N3O3S | 10MG
SDP
Catalog No. 5000261681
Encompass_Preferred
Lp-PLA2-IN-3 is a potent, orally bioavailable small-molecule inhibitor of lipoprotein-associated phospholipase A2 used for biochemical and preclinical research. It exhibits an IC50 of 14 nM against recombinant human Lp-PLA2 and is supplied at high purity for assay and pharmacology applications.
  • Potent inhibition: IC50 of 14 nM against recombinant human Lp-PLA2.
  • Orally bioavailable small molecule suitable for in vivo studies.
  • High purity (≈99.9%) for consistent experimental results.
  • Provided in multiple quantities and solution formats for flexibility.
  • Soluble in DMSO; recommended for preparing stock solutions.
  • Appropriate for target validation, enzymology, and pharmacology studies.
Catalog No. 50-002-61681 Supplier Medchemexpress LLC Supplier No. HY13314910MG
Lp-PLA2-IN-3 is a potent, orally bioavailable small-molecule inhibitor of lipoprotein-associated phospholipase A2 used for biochemical and preclinical research. It exhibits an IC50 of 14 nM against recombinant human Lp-PLA2 and is supplied at high purity for assay and pharmacology applications.
  • Potent inhibition: IC50 of 14 nM against recombinant human Lp-PLA2.
  • Orally bioavailable small molecule suitable for in vivo studies.
  • High purity (≈99.9%) for consistent experimental results.
  • Provided in multiple quantities and solution formats for flexibility.
  • Soluble in DMSO; recommended for preparing stock solutions.
  • Appropriate for target validation, enzymology, and pharmacology studies.