Learn More
MSC BluShield Soap Tip, 65 Degree, Black, Low Pressure, Quick Disconnect
Catalog No. 502838084
Encompass
This soap tip is designed for efficient low-pressure soap application with pressure washers, ensuring effective and even coverage. It features a 65-degree spray angle and a quick disconnect design for easy attachment and removal.
  • Designed for applying soap at low pressure, ensuring effective and even coverage for cleaning tasks.
  • Features a 65-degree spray angle and large orifice, providing a wide and consistent low-pressure spray pattern.
  • Easily connects to pressure washer wands equipped with a 1/4 in quick disconnect socket.
  • Allows for quick and easy attachment and removal, enabling swift changes between different spray tips.
  • Black color coding ensures easy identification.
Catalog No. 50-283-8084 Supplier MSC Supplier No. 63956395
This soap tip is designed for efficient low-pressure soap application with pressure washers, ensuring effective and even coverage. It features a 65-degree spray angle and a quick disconnect design for easy attachment and removal.
  • Designed for applying soap at low pressure, ensuring effective and even coverage for cleaning tasks.
  • Features a 65-degree spray angle and large orifice, providing a wide and consistent low-pressure spray pattern.
  • Easily connects to pressure washer wands equipped with a 1/4 in quick disconnect socket.
  • Allows for quick and easy attachment and removal, enabling swift changes between different spray tips.
  • Black color coding ensures easy identification.