Learn More
Grainger Vestil Manual Cylinder Lifter, Steel, Powder Coated, 200 lb. Capacity, 1 ea
Catalog No. 502775041
Encompass_Preferred
The LP Tank Lifter is a manual lifting solution designed for use in manufacturing and maintenance facilities. It is constructed from powder-coated steel to provide durability during daily operations. The unit features a bright yellow finish for increased visibility and safety in indoor and outdoor settings. It operates by attaching to a lifting device via a centrally located hook opening, allowing the arms to secure the LP tank at the top and base ridges for transport.
  • Capacity of 200 lb.
  • Accommodates tank diameters of 25 in. to 30 in.
  • Constructed from heavy-duty steel
  • Includes two swivel hooks
  • Features a compact and vertical design for efficient handling
  • Assembled dimensions of 34 5/16 in. W x 7/8 in. D x 15 9/16 in. H
Catalog No. 50-277-5041 Supplier Grainger Supplier No. 172M01
The LP Tank Lifter is a manual lifting solution designed for use in manufacturing and maintenance facilities. It is constructed from powder-coated steel to provide durability during daily operations. The unit features a bright yellow finish for increased visibility and safety in indoor and outdoor settings. It operates by attaching to a lifting device via a centrally located hook opening, allowing the arms to secure the LP tank at the top and base ridges for transport.
  • Capacity of 200 lb.
  • Accommodates tank diameters of 25 in. to 30 in.
  • Constructed from heavy-duty steel
  • Includes two swivel hooks
  • Features a compact and vertical design for efficient handling
  • Assembled dimensions of 34 5/16 in. W x 7/8 in. D x 15 9/16 in. H