Learn More
Eicom USA LP-TUBE 0.5 100/CS
Catalog No. NC0503158
Encompass
Microresico Low Bind 0.5 ml Tube. Low Protein Binding, Low Elution, High Fluid Recovery, Rated to 16,000 g, Frosted flat cap for easy writing Autoclavable (121 deg.C for 20 min.) 100 tubes/pack
Catalog No. NC0503158 Supplier Eicom USA Supplier No. RC9.201.60
Microresico Low Bind 0.5 ml Tube. Low Protein Binding, Low Elution, High Fluid Recovery, Rated to 16,000 g, Frosted flat cap for easy writing Autoclavable (121 deg.C for 20 min.) 100 tubes/pack