Learn More
Medchemexpress LLC LP17 TFA (LQVTDSGLYRCVIYHPP TFA) | 99.2% | 1961.24 | 5 MG
SDP
Catalog No. 5000398280
Encompass_Preferred
LP17 (LQVTDSGLYRCVIYHPP) TFA is a BBB-penetrable triggering receptor expressed on myeloid cells (TREM-1) inhibitory peptide. It substantially alleviates ischemia-induced infarction and neuronal injury by accessing the brain and blocking TREM-1. For research use only.
  • Decreases mRNA levels of pro-inflammatory cytokines and chemokines.
  • Attenuates extracellular protein levels of IL-1β and IL-18 in a microglia oxygen-glucose deprivation (OGD) model.
  • Interacts with microglial SYK.
  • Alleviates ischemia-induced infarction and neuronal injury in mice.
  • Reduces infarct volume by 27.3%.
  • Reduces TUNEL positive cells and FJC positive neurons.
  • Rescues neurological deficits and cognitive dysfunction.
  • Inhibits microglial M1 polarization and neutrophil infiltration.
Catalog No. 50-003-98280 Supplier Medchemexpress LLC Supplier No. HYP3400A5MG
LP17 (LQVTDSGLYRCVIYHPP) TFA is a BBB-penetrable triggering receptor expressed on myeloid cells (TREM-1) inhibitory peptide. It substantially alleviates ischemia-induced infarction and neuronal injury by accessing the brain and blocking TREM-1. For research use only.
  • Decreases mRNA levels of pro-inflammatory cytokines and chemokines.
  • Attenuates extracellular protein levels of IL-1β and IL-18 in a microglia oxygen-glucose deprivation (OGD) model.
  • Interacts with microglial SYK.
  • Alleviates ischemia-induced infarction and neuronal injury in mice.
  • Reduces infarct volume by 27.3%.
  • Reduces TUNEL positive cells and FJC positive neurons.
  • Rescues neurological deficits and cognitive dysfunction.
  • Inhibits microglial M1 polarization and neutrophil infiltration.