Learn More
Echelon Biosciences Research Labs XY-17 (LPA3 Agonist) 0.5 mg
SDP
Catalog No. 50720527
Encompass
XY-17 is a fluorinated phosphonate analog of lysophosphatidic acid (LPA). XY-17 is not hydrolyzed by lipid phosphate phosphatase (LPP) and acts as an agonist of the LPA3 receptor. This item is non returnable
This item is not returnable. View return policy
Catalog No. 50-720-527 Supplier Echelon Biosciences Research Labs Supplier No. L9118500UG
XY-17 is a fluorinated phosphonate analog of lysophosphatidic acid (LPA). XY-17 is not hydrolyzed by lipid phosphate phosphatase (LPP) and acts as an agonist of the LPA3 receptor. This item is non returnable