Learn More
Echelon Biosciences Research Labs XY-14 (Lipid Phosphate Phosphatase 1 Inhibitor) 1 mg
SDP
Supplier:  Echelon Biosciences Research Labs L92181MG
Encompass
XY-14 is a competitive inhibitor of Lipid Phosphate Phosphatase 1 (LPP1) which is the major enzyme responsible for degrading lysophosphatidic acid (LPA) in platelets. XY-14 (10 M) inhibits LPP1 in human pulmonary artery endothelial cells, preventing the dephosphorylation of sphingosine 1-phosphate (S1P) to sphingosine. XY-14 is also reported to inhibit LPP2 & LPP3 This item is non returnable
Catalog No. 50-720-529