Learn More
Echelon Biosciences Research Labs Lysophosphatidic Acid ELISA Kit
SDP
Supplier:  Echelon Biosciences Research Labs K2800SEAKIT
Encompass
The Lysophosphatidic Acid ELISA kit (LPA Assay Kit II) is a sensitive and robust method for quantification of serum Lysophosphatidic Acid (LPA) in a 96-well plate competitive ELISA format.Sample Types: Serum and Plasma (human or other animal)Sample Volume: 20 L/sample (for duplicate points)Assay Incubation Time: 2 hours and 30 minutesAssay Range: 0.064 M to 1,000 M This item is non returnable
Catalog No. 50-822-52