Learn More
Echelon Biosciences Research Labs LPA Beads 1 mL
SDP
Supplier:  Echelon Biosciences Research Labs L61011ML
Encompass
LPA beads are composed of agarose with 10 nanomoles of bound Lyosphosphatidic Acid (LPA) per mL of beads, enough for several protein binding experiments. Each 1 mL order of LPA Beads includes a small amount of control beads. Larger quantities of Control Beads are also available for purchase. This item is non returnable
Catalog No. 50-720-522