Learn More
Echelon Biosciences Research Labs LPA-BIOTIN 50 ug
SDP
Supplier:  Echelon Biosciences Research Labs L012B50UG
Encompass
Lysophosphatidic Acid (LPA) is a small lysophospholipid involved in diverse cellular processes such as cell proliferation, chemotaxis, platelet aggregation, wound healing, angiogenesis, tumor invasion, and smooth muscle contraction. LPA binds to several G-coupled protein receptors to initiate its biological functions. In cancer, LPA primarily promotes cell survival, migration and invasion. LPA Biotin is labeled at the sn-1 position. This item is non returnable
Catalog No. 50-720-469