Learn More
Eurofins Discoverx Ready-to-Assay LPA5/GPR92 Lysophospholipid Receptor Frozen Cells, 2 Vials of Cells. (HTS209RTA)
Catalog No. 502006058
Encompass
The Ready-to-Assay LPA5/GPR92 Lysophospholipid Receptor Frozen Cells are a simple, thaw-and-assay cell format for use in studying drug candidates (small molecules or biologics) targeting GPCRs. This assay assesses ligand (e.g. LPA) based activation of LPAR5 GPCR activity via detection of increasing levels of intracellular calcium. The included cell line overexpresses LPAR5 alongside endogenous high levels of a promiscuous G-protein, coupling the receptor to the calcium signaling pathway. Upon activation of LPAR5, the resulting calcium activity can be fluorescently assayed. Ready-to-assay kits consist of two vials of the stably modified clonal cell line in a convenient thaw-and-use format. For detailed product information, User manual and datasheet, visit www.discoverx.com/products-applications/frozen-cells.
Catalog No. 50-200-6058 Supplier Eurofins Discoverx Supplier No. HTS209RTA
The Ready-to-Assay LPA5/GPR92 Lysophospholipid Receptor Frozen Cells are a simple, thaw-and-assay cell format for use in studying drug candidates (small molecules or biologics) targeting GPCRs. This assay assesses ligand (e.g. LPA) based activation of LPAR5 GPCR activity via detection of increasing levels of intracellular calcium. The included cell line overexpresses LPAR5 alongside endogenous high levels of a promiscuous G-protein, coupling the receptor to the calcium signaling pathway. Upon activation of LPAR5, the resulting calcium activity can be fluorescently assayed. Ready-to-assay kits consist of two vials of the stably modified clonal cell line in a convenient thaw-and-use format. For detailed product information, User manual and datasheet, visit www.discoverx.com/products-applications/frozen-cells.