Learn More
Bioworld LPALINEAR POLYACRYLAMIDE)
Supplier:  Bioworld 105100001
Encompass_Preferred
LPA (LINEAR POLYACRYLAMIDE) FOR DNA/RNA PRECIPITATION: LPA is an inert coprecipitant used to aid recovery of nucleic acids during alcohol precipitations, essential for quantitative recovery of small amounts of nucleic acids in dilute sol...
Catalog No. 50-196-648